Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromo-1,2,3,4-tetrahydro-1,8-naphthyridine
6-Bromo-1,2,3,4-tetrahydro-1,8-naphthyridine
6-Bromo-1,2,3,4-tetrahydro-1,8-naphthyridine
:
Apollo Scientific
6-Bromo-1,2,3,4-tetrahydro-1,8-naphthyridine
:
Apollo Scientific
:
1023813-80-0
:
:
C₈H₉BrN₂
:
MFCD09881254
:
213.074 g/mol
:
GHS07
:
Ambient