
(4-Oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17547-79-4
:
Formula:C₁₁H₁₁NO₃S
MDL Number:MFCD01416244
:237.275 g/mol
Melting Pt:225…230 °C
:GHS07
Storage Temperature:Ambient