Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-4-Benzyl-1,3-thiazolidine-2-thione
(R)-4-Benzyl-1,3-thiazolidine-2-thione 98%
(R)-4-Benzyl-1,3-thiazolidine-2-thione 98%
Irritant
(R)-4-Benzyl-1,3-thiazolidine-2-thione 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
(R)-4-Benzyl-1,3-thiazolidine-2-thione 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
110199-17-2
:
Formula:
C₁₀H₁₁NS₂
MDL Number:
MFCD06658217
:
209.331 g/mol
:
GHS07
Storage Temperature:
Ambient