Order Entry
Export
ContactUsLinkComponent
4-(4-Bromophenyl)-2-methyl-1,3-thiazole 97+%
Irritant
4-(4-Bromophenyl)-2-methyl-1,3-thiazole 97+%
Catalog # APOSOR1423-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(4-Bromophenyl)-2-methyl-1,3-thiazole 97+%
Catalog # APOSOR1423-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :254.146 g/mol
Melting Pt:130…137 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:66047-74-3
Formula:C₁₀H₈BrNS
MDL Number:MFCD00098207