Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(3,4-Dichlorobenzyl)piperidine hydrochloride
4-(3,4-Dichlorobenzyl)piperidine hydrochloride
4-(3,4-Dichlorobenzyl)piperidine hydrochloride
4-(3,4-Dichlorobenzyl)piperidine hydrochloride
APOSOR12873-1G
:
Apollo Scientific
:
4-(3,4-Dichlorobenzyl)piperidine hydrochloride
APOSOR12873-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
280.621 g/mol
:
GHS07
:
Ambient
:
1171138-69-4
:
C₁₂H₁₅Cl₂N·ClH
:
MFCD03840139