Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dichloroquinoline-3-carboxaldehyde
2,6-Dichloroquinoline-3-carboxaldehyde
2,6-Dichloroquinoline-3-carboxaldehyde
Irritant
2,6-Dichloroquinoline-3-carboxaldehyde
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,6-Dichloroquinoline-3-carboxaldehyde
Supplier:
Apollo Scientific
About this item
CAS Number:
73568-41-9
:
Formula:
C₁₀H₅Cl₂NO
MDL Number:
MFCD06202347
:
226.059 g/mol
Melting Pt:
191…192 °C (dec.)
:
GHS07
Storage Temperature:
Ambient