Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(3,5-Dichlorophenyl)pyrrolidine
2-(3,5-Dichlorophenyl)pyrrolidine
2-(3,5-Dichlorophenyl)pyrrolidine
APOSOR12506-250MG
:
Apollo Scientific
:
2-(3,5-Dichlorophenyl)pyrrolidine
APOSOR12506-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
216.107 g/mol
:
GHS07
:
Refrigerator
:
887344-13-0
:
C₁₀H₁₁Cl₂N
:
MFCD02684093