0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-(3,4-Dichlorophenoxy)aniline
4-(3,4-Dichlorophenoxy)aniline
4-(3,4-Dichlorophenoxy)aniline
APOSOR11352-5G
:
Apollo Scientific
:
4-(3,4-Dichlorophenoxy)aniline
APOSOR11352-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
254.112 g/mol
:
68.7…70.3 °C
:
GHS07
:
Refrigerator
:
67651-53-0
:
C₁₂H₉Cl₂NO
:
MFCD00459613