N-(4-Imidazo[1,2-a]pyrimidin-2-ylphenyl)acetamide
Catalog # APOSOR111615-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:252.271 g/mol
Storage Temperature:Ambient
CAS Number:103779-36-8
Formula:C₁₄H₁₂N₄O
MDL Number:MFCD05713256