2-(2,4-Dichlorophenoxy)nicotinonitrile
Catalog # APOSOR111487-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:265.095 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₆Cl₂N₂O
MDL Number:MFCD00277879