
7-Methylimidazo[1,2-a]pyridine-8-carbonitrile
Catalog # APOSOR111479-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:157.172 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1352888-45-9
Formula:C₉H₇N₃
MDL Number:MFCD22553043