Order Entry
Export
ContactUsLinkComponent
3,4-Dichloro-N'-[imidazo[1,2-a]pyridin-6-ylmethylene]benzohydrazide
3,4-Dichloro-N'-[imidazo[1,2-a]pyridin-6-ylmethylene]benzohydrazide
Catalog # APOSOR111476-1G
Supplier:  Apollo Scientific
CAS Number:  
3,4-Dichloro-N'-[imidazo[1,2-a]pyridin-6-ylmethylene]benzohydrazide
Catalog # APOSOR111476-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :333.172 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₀Cl₂N₄O
MDL Number:MFCD31380611