6-Chloro-3-nitroimidazo[1,2-a]pyridine
Catalog # APOSOR111419-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:197.579 g/mol
Storage Temperature:Ambient
CAS Number:25045-84-5
Formula:C₇H₄ClN₃O₂
MDL Number:MFCD25961521