Order Entry
Export
ContactUsLinkComponent
2-(Chloromethyl)-6-methoxy-1,3-benzoxazole
Irritant
2-(Chloromethyl)-6-methoxy-1,3-benzoxazole
Catalog # APOSOR111389-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Chloromethyl)-6-methoxy-1,3-benzoxazole
Catalog # APOSOR111389-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :197.618 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:944897-67-0
Formula:C₉H₈ClNO₂
MDL Number:MFCD10696616