Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Cyclopropylthieno[2,3-d]pyrimidin-4-amine
N-Cyclopropylthieno[2,3-d]pyrimidin-4-amine
N-Cyclopropylthieno[2,3-d]pyrimidin-4-amine
N-Cyclopropylthieno[2,3-d]pyrimidin-4-amine
APOSOR111366-1G
:
Apollo Scientific
:
N-Cyclopropylthieno[2,3-d]pyrimidin-4-amine
APOSOR111366-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
191.253 g/mol
:
Ambient
:
63893-75-4
:
C₉H₉N₃S
:
MFCD11176831