(1,3-Thiazol-2-ylmethylene)malononitrile
Catalog # APOSOR111279-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:161.184 g/mol
Storage Temperature:Ambient
CAS Number:771565-96-9
Formula:C₇H₃N₃S
MDL Number:MFCD06214304