
3,7-Dichloroimidazo[1,2-a]pyridine
Catalog # APOSOR111274-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:187.026 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1019026-71-1
Formula:C₇H₄Cl₂N₂
MDL Number:MFCD09994785