
3,6-Dichloroimidazo[1,2-a]pyridine
Catalog # APOSOR111273-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:187.026 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1019027-83-8
Formula:C₇H₄Cl₂N₂
MDL Number:MFCD09994777