
Imidazo[1,2-a]pyridine-6-carbaldehyde oxime
Catalog # APOSOR111254-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:161.161 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1709825-51-3
Formula:C₈H₇N₃O
MDL Number:MFCD30188177