
Methyl 4-(2,2-dicyanovinyl)benzoate
Catalog # APOSOR111251-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:212.204 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
CAS Number:3129-16-6
Formula:C₁₂H₈N₂O₂
MDL Number:MFCD01141799