Order Entry
Export
ContactUsLinkComponent
4-(5-Oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)benzoic acid
4-(5-Oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)benzoic acid
Catalog # APOSOR111220-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(5-Oxo-1,5-dihydro-4H-1,2,4-triazol-4-yl)benzoic acid
Catalog # APOSOR111220-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :205.17 g/mol
Storage Temperature:Ambient
Formula:C₉H₇N₃O₃
MDL Number:MFCD29918595