
Ethyl imidazo[1,2-a]pyridin-2-ylacetate
Catalog # APOSOR111219-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.225 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:21755-34-0
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD01113258