
5-Methoxy-N-methyl-1,3-benzoxazol-2-amine
Catalog # APOSOR111206-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:178.188 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1267987-80-3
Formula:C₉H₁₀N₂O₂
MDL Number:MFCD19374710