Order Entry
Export
ContactUsLinkComponent
5-Chloro-2-propyl-1,3-benzoxazole
5-Chloro-2-propyl-1,3-benzoxazole
Catalog # APOSOR111198-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Chloro-2-propyl-1,3-benzoxazole
Catalog # APOSOR111198-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.645 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₀ClNO
MDL Number:MFCD19375141