4-(3-Bromoimidazo[1,2-a]pyridin-2-yl)benzonitrile
Catalog # APOSOR111171-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:298.137 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₈BrN₃
MDL Number:MFCD28992258