Order Entry
Export
ContactUsLinkComponent
4-Imidazo[1,2-a]pyridin-2-ylbenzonitrile
Irritant
4-Imidazo[1,2-a]pyridin-2-ylbenzonitrile
Catalog # APOSOR111163-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Imidazo[1,2-a]pyridin-2-ylbenzonitrile
Catalog # APOSOR111163-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.241 g/mol
Melting Pt:211…215 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:55843-91-9
Formula:C₁₄H₉N₃
MDL Number:MFCD05863434