Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Imidazo[1,2-a]pyridine-8-carbonitrile
Imidazo[1,2-a]pyridine-8-carbonitrile
Imidazo[1,2-a]pyridine-8-carbonitrile
:
Apollo Scientific
Imidazo[1,2-a]pyridine-8-carbonitrile
:
Apollo Scientific
:
136117-70-9
:
:
C₈H₅N₃
:
MFCD09994637
:
143.145 g/mol
:
GHS07
:
Ambient