
4-tert-Butyl-1,3-thiazole-2-carboxaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:211942-98-2
:
Formula:C₈H₁₁NOS
MDL Number:MFCD07381826
:169.244 g/mol
Boiling Pt:50…52 °C (2 mm Hg)
:GHS07
Storage Temperature:Ambient