N,5-Dimethylthiophene-2-sulphonamide
Catalog # APOSOR111074-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.271 g/mol
Storage Temperature:Ambient
CAS Number:61714-51-0
Formula:C₆H₉NO₂S₂
MDL Number:MFCD11171682