Thieno[3,2-e][1,2,4]triazolo[4,3-c]pyrimidine
Catalog # APOSOR110985-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:176.198 g/mol
Storage Temperature:Ambient
CAS Number:18740-36-8
Formula:C₇H₄N₄S
MDL Number:MFCD26939174