
2-(3,4-Dichlorophenyl)acetamide
Catalog # APOSOR110962-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.053 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:868697-78-3
Formula:C₈H₇Cl₂NO
MDL Number:MFCD11043647