Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(3,4-Dimethoxyphenyl)ethanol
1-(3,4-Dimethoxyphenyl)ethanol
1-(3,4-Dimethoxyphenyl)ethanol
1-(3,4-Dimethoxyphenyl)ethanol
Supplier:
Apollo Scientific
Certificates
Expand image
1-(3,4-Dimethoxyphenyl)ethanol
Supplier:
Apollo Scientific
About this item
CAS Number:
5653-65-6
:
Formula:
C₁₀H₁₄O₃
MDL Number:
MFCD02127273
:
182.216 g/mol
Storage Temperature:
Ambient