Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 3-(4,5-dimethoxy-2-nitrophenyl)acrylate
Methyl 3-(4,5-dimethoxy-2-nitrophenyl)acrylate
Methyl 3-(4,5-dimethoxy-2-nitrophenyl)acrylate
Methyl 3-(4,5-dimethoxy-2-nitrophenyl)acrylate
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 3-(4,5-dimethoxy-2-nitrophenyl)acrylate
Supplier:
Apollo Scientific
About this item
CAS Number:
118509-96-9
:
Formula:
C₁₂H₁₃NO₆
MDL Number:
MFCD00266189
:
267.235 g/mol
Storage Temperature:
Ambient