5-Methoxy-2-methyl-1,3-benzoxazole
Catalog # APOSOR110870-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:163.173 g/mol
Storage Temperature:Ambient
CAS Number:5676-57-3
Formula:C₉H₉NO₂
MDL Number:MFCD00128831