Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dinitro-1,3-benzodioxole
5,6-Dinitro-1,3-benzodioxole 95%
5,6-Dinitro-1,3-benzodioxole 95%
Irritant
5,6-Dinitro-1,3-benzodioxole 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5,6-Dinitro-1,3-benzodioxole 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
7748-59-6
:
Formula:
C₇H₄N₂O₆
MDL Number:
MFCD04973596
:
212.116 g/mol
:
GHS07
Storage Temperature:
Ambient