
3-(1,1-Dioxidothiomorpholin-4-yl)propanoic acid
Catalog # APOSOR110858-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.247 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:849928-19-4
Formula:C₇H₁₃NO₄S
MDL Number:MFCD03787455