Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
3-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
3-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
APOSOR110856-1G
:
Apollo Scientific
:
3-(1,1-Dioxidothiomorpholin-4-yl)benzoic acid
APOSOR110856-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
255.29 g/mol
:
215…217 °C
:
Ambient
:
763073-96-7
:
C₁₁H₁₃NO₄S
:
MFCD06659598