

3-Bromo-5-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Catalog # APOSOR110724-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:275.487 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:1227955-02-3
Formula:C₈H₄BrClN₂O₂
MDL Number:MFCD11846272