Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Phenyl-2-azaspiro[4.4]nonane-1,3-dione
2-Phenyl-2-azaspiro[4.4]nonane-1,3-dione
2-Phenyl-2-azaspiro[4.4]nonane-1,3-dione
2-Phenyl-2-azaspiro[4.4]nonane-1,3-dione
Supplier:
Apollo Scientific
Certificates
Expand image
2-Phenyl-2-azaspiro[4.4]nonane-1,3-dione
Supplier:
Apollo Scientific
About this item
CAS Number:
856068-76-3
:
Formula:
C₁₄H₁₅NO₂
MDL Number:
MFCD20037419
:
229.274 g/mol
Storage Temperature:
Ambient