5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carbaldehyde
Catalog # APOSOR110638-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:152.214 g/mol
Storage Temperature:Ambient
CAS Number:956010-59-6
Formula:C₈H₈OS
MDL Number:MFCD17276640