Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dimethyl-1,3-thiazole-5-sulphonamide
2,4-Dimethyl-1,3-thiazole-5-sulphonamide
2,4-Dimethyl-1,3-thiazole-5-sulphonamide
Irritant
2,4-Dimethyl-1,3-thiazole-5-sulphonamide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Dimethyl-1,3-thiazole-5-sulphonamide
Supplier:
Apollo Scientific
About this item
CAS Number:
80466-90-6
:
Formula:
C₅H₈N₂O₂S₂
MDL Number:
MFCD00178766
:
192.259 g/mol
:
GHS07
Storage Temperature:
Refrigerator