1-Methyl-4-(3-methyl-4-nitrophenyl)piperazine
Catalog # APOSOR110482-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.282 g/mol
Storage Temperature:Ambient
CAS Number:16154-61-3
Formula:C₁₂H₁₇N₃O₂
MDL Number:MFCD11039716