0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Methyl 2,3-dimethylquinoxaline-6-carboxylate
Methyl 2,3-dimethylquinoxaline-6-carboxylate
Methyl 2,3-dimethylquinoxaline-6-carboxylate
APOSOR110390-1G
:
Apollo Scientific
:
Methyl 2,3-dimethylquinoxaline-6-carboxylate
APOSOR110390-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
216.236 g/mol
:
GHS07
:
Ambient
:
32461-66-8
:
C₁₂H₁₂N₂O₂
:
MFCD00205437