Order Entry
Export
ContactUsLinkComponent
2-(2-Methyl-1,3-thiazol-4-yl)acetohydrazide
Irritant
2-(2-Methyl-1,3-thiazol-4-yl)acetohydrazide
Catalog # APOSOR110344-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(2-Methyl-1,3-thiazol-4-yl)acetohydrazide
Catalog # APOSOR110344-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :171.22 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:496057-29-5
Formula:C₆H₉N₃OS
MDL Number:MFCD03789239