2-Propyl[1,3]oxazolo[4,5-b]pyridine
Catalog # APOSOR110331-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:162.189 g/mol
Storage Temperature:Ambient
CAS Number:104711-71-9
Formula:C₉H₁₀N₂O
MDL Number:MFCD07021391