
Methyl 2-ethoxy-5-iodonicotinate
Catalog # APOSOR110282-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:307.085 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:335078-24-5
Formula:C₉H₁₀INO₃
MDL Number:MFCD13194591