Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dichloro-2-propyl-1H-benzimidazole
5,6-Dichloro-2-propyl-1H-benzimidazole
5,6-Dichloro-2-propyl-1H-benzimidazole
5,6-Dichloro-2-propyl-1H-benzimidazole
Supplier:
Apollo Scientific
Certificates
Expand image
5,6-Dichloro-2-propyl-1H-benzimidazole
Supplier:
Apollo Scientific
About this item
CAS Number:
176949-20-5
:
Formula:
C₁₀H₁₀Cl₂N₂
MDL Number:
MFCD08064758
:
229.106 g/mol
Storage Temperature:
Ambient