
3-Iodoimidazo[1,2-a]pyridine-6-carbonitrile
Catalog # APOSOR110208-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.042 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:885276-13-1
Formula:C₈H₄IN₃
MDL Number:MFCD06739236