
3-Chloroimidazo[1,2-a]pyridine-6-carbonitrile
Catalog # APOSOR110191-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:177.59 g/mol
Melting Pt:199…201 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1019027-68-9
Formula:C₈H₄ClN₃
MDL Number:MFCD09994775