0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
4-Methyl-2-phenyl-1,3-oxazole
4-Methyl-2-phenyl-1,3-oxazole
4-Methyl-2-phenyl-1,3-oxazole
4-Methyl-2-phenyl-1,3-oxazole
APOSOR110144-1G
:
Apollo Scientific
:
4-Methyl-2-phenyl-1,3-oxazole
APOSOR110144-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
159.185 g/mol
:
60 °C (1 mm Hg)
:
GHS07
:
Ambient
:
877-39-4
:
C₁₀H₉NO
:
MFCD11055268